Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002966.2 | NP_002957.1 | 3 | 749 | ||
GenBank mRNA | AK291073.1 | - | 3 | 341 | ||
BC015973.1 | - | 3 | 343 | 4103596 | ||
BC105786.1 | - | 3 | 327 | 6701084 | ||
BQ050753.1 | - | 3 | 334 | |||
CR542162.1 | - | 2 | 241 | |||
M38591.1 | - | 3 | 352 | |||
M81457.1 | - | 3 | 312 |