Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010001.2 | NP_034131.2 | 3 | 486 | ||
GenBank mRNA | AF047542.1 | - | 470 | |||
AK005017.1 | - | 3 | 487 | |||
BC057912.1 | - | 471 | 5052873 |
Gene Symbol: Cyp2c50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167875.1 | NP_001161347.1 | 3 | 491 | ||
NM_001167877.1 | NP_001161349.1 | 3 | 491 | |||
NM_134144.2 | NP_598905.2 | 3 | 491 | |||
GenBank mRNA | AK149466.1 | - | 3 | 436 | ||
AY206873.1 | - | 3 | 470 | |||
BC011222.1 | - | 3 | 474 | 4195223 | ||
BC051050.1 | - | 3 | 482 | 5052536 |
Gene Symbol: Cyp2c54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206537.2 | NP_996260.1 | 3 | 472 | ||
GenBank mRNA | AI181879.1 | - | 3 | 461 | ||
BC043929.1 | - | 2 | 254 |