Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146200.1 | NP_666312.1 | 3 | 186 | ||
GenBank mRNA | AK009925.1 | - | 3 | 226 | ||
AK013160.1 | - | 3 | 213 | |||
AK076165.1 | - | 3 | 204 | |||
AK088076.1 | - | 3 | 229 | |||
AK152693.1 | - | 3 | 204 | |||
AK160078.1 | - | 3 | 206 | |||
AK166743.1 | - | 3 | 229 | |||
AK168160.1 | - | 3 | 191 | |||
AK168174.1 | - | 3 | 191 | |||
BC024855.1 | - | 3 | 186 | 5359060 | ||
BC025032.1 | - | 3 | 207 | 4947435 |