Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146609.2 | NP_666820.2 | 1 | 461 |
Set Membership: |
G-protein coupled receptor |