Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA2-SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199744.1 | NP_001186673.1 | 5 | 495 | ||
GenBank mRNA | BG616641.1 | - | 5 | 457 | ||
BX117371.1 | - | 5 | 440 |
Gene Symbol: SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006512.3 | NP_006503.2 | 3 | 310 | ||
GenBank mRNA | AK312182.1 | - | 3 | 261 | ||
AV699045.1 | - | 3 | 253 | |||
BC007026.1 | - | 3 | 242 | 4071711 | ||
BG569210.1 | - | 3 | 311 | |||
CB113387.1 | - | 3 | 283 | |||
CR541758.1 | - | 2 | 185 | |||
M81349.1 | - | 3 | 260 |