Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 7 | 982 | ||
NM_001164488.1 | NP_001157960.1 | 7 | 915 | |||
NM_011710.3 | NP_035840.3 | 7 | 928 | |||
GenBank mRNA | AK004541.1 | - | 7 | 916 | ||
AK007754.1 | - | 5 | 544 | |||
AK150090.1 | - | 8 | 946 | |||
AK150267.1 | - | 8 | 1030 | |||
AK152966.1 | - | 7 | 946 | |||
AK153337.1 | - | 7 | 953 | |||
BC003450.1 | - | 7 | 914 | 2654231 | ||
BC046232.1 | - | 7 | 921 | 6813406 | ||
CF731228.1 | - | 4 | 434 | |||
CT010258.1 | - | 6 | 814 |