Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164314.1 | NP_001157786.1 | 7, 8 | 1009 | ||
NM_001164488.1 | NP_001157960.1 | 7, 8 | 942 | |||
NM_011710.3 | NP_035840.3 | 7, 8 | 955 | |||
GenBank mRNA | AK004541.1 | - | 7, 8 | 943 | ||
AK007754.1 | - | 5, 6 | 571 | |||
AK150090.1 | - | 8, 9 | 973 | |||
AK150267.1 | - | 8, 9 | 1057 | |||
AK152966.1 | - | 7, 8 | 973 | |||
AK153337.1 | - | 7, 8 | 980 | |||
BC003450.1 | - | 7, 8 | 941 | 2654231 | ||
BC046232.1 | - | 7, 8 | 948 | 6813406 | ||
CF731228.1 | - | 4, 5 | 461 | |||
CT010258.1 | - | 6, 7 | 841 |