Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005622.3 | NP_005613.2 | 9 | 1347 | ||
NM_202000.2 | NP_973729.1 | 9 | 1347 | |||
XM_011545911.1 | XP_011544213.1 | 10 | 1452 | |||
XM_017023523.1 | XP_016879012.1 | 9 | 1341 | |||
XM_017023524.1 | XP_016879013.1 | 8 | 1211 | |||
GenBank mRNA | AK223139.1 | - | 9 | 1316 | ||
AK290322.1 | - | 9 | 1316 | |||
AK296122.1 | - | 9 | 1146 | |||
BC002790.2 | - | 8 | 1206 | 3617826 | ||
BI826935.1 | - | 3 | 222 | |||
BU170035.1 | - | 3 | 199 | |||
D16350.1 | - | 8 | 1187 | |||
X80062.1 | - | 9 | 1296 |