Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005622.3 | NP_005613.2 | 6 | 998 | ||
NM_202000.2 | NP_973729.1 | 6 | 998 | |||
XM_011545911.1 | XP_011544213.1 | 7 | 1103 | |||
XM_017023523.1 | XP_016879012.1 | 6 | 992 | |||
XM_017023524.1 | XP_016879013.1 | 5 | 862 | |||
GenBank mRNA | AK223139.1 | - | 6 | 967 | ||
AK290322.1 | - | 6 | 967 | |||
AK296122.1 | - | 6 | 797 | |||
BC002790.2 | - | 5 | 857 | 3617826 | ||
D16350.1 | - | 5 | 838 | |||
X80062.1 | - | 6 | 947 |