Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3f3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008210.4 | NP_032236.1 | 4 | 967 | ||
GenBank mRNA | AI155695.1 | - | 1 | 44 | ||
AK037900.1 | - | 4 | 969 | |||
AK142749.1 | - | 1 | 191 | |||
AK150928.1 | - | 4 | 922 | |||
AK152293.1 | - | 4 | 922 | |||
AK160677.1 | - | 4 | 926 | |||
AK168197.1 | - | 4 | 926 | |||
AK169608.1 | - | 4 | 922 | |||
BC002268.1 | - | 4 | 912 | 3495830 | ||
BC012687.1 | - | 4 | 916 | 4224280 | ||
BC083353.1 | - | 4 | 907 | 5714859 | ||
BC088835.1 | - | 4 | 904 | 6437687 | ||
BC106177.1 | - | 4 | 900 | 6511787 |
Gene Symbol: Gm12657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081019.1 | NP_001074488.1 | 2 | 856 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC027275.1 | - | 2442 | Chr. - - [-] on Build |