Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303252.1 | NP_001290181.1 | 4 | 608 | ||
NM_002969.4 | NP_002960.2 | 4 | 608 | |||
GenBank mRNA | AK307421.1 | - | 3 | 196 | ||
BC015741.1 | - | 4 | 509 | 4907996 | ||
BX640648.1 | - | 3 | 245 | |||
CB123561.1 | - | 3 | 181 | |||
CR456515.1 | - | 4 | 393 | |||
U66243.1 | - | 4 | 760 | |||
X79483.1 | - | 4 | 391 | |||
Y10487.1 | - | 4 | 564 |