Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 6 | 605 | ||
NR_027783.2 | 7 | 715 | ||||
XM_017029720.1 | XP_016885209.1 | 3 | 1233 | |||
GenBank mRNA | AF251292.1 | - | 6 | 628 | ||
AK312162.1 | - | 6 | 588 | |||
AL050290.1 | - | 7 | 676 | |||
AY841998.1 | - | 7 | 522 | |||
BC002503.2 | - | 6 | 566 | 3051095 | ||
BC008424.1 | - | 6 | 590 | 4134501 | ||
BT006825.1 | - | 6 | 411 | |||
CD516700.1 | - | 6 | 611 | |||
CR450294.1 | - | 6 | 411 | |||
M55580.1 | - | 416 | ||||
M77693.1 | - | 6 | 576 |