Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm7325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177468.1 | NP_001170939.1 | 2 | 678 | ||
NM_001177469.1 | NP_001170940.1 | 2 | 681 | |||
NM_001177470.1 | NP_001170941.1 | 2 | 722 | |||
GenBank mRNA | BB399187.1 | - | 1 | 142 | ||
CJ191178.1 | - | 1 | 416 | |||
DQ190000.1 | - | 2 | 677 |