Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF218078.1 | - | 2 | 240 | ||
AF218079.1 | - | 2 | 240 | |||
AY860973.1 | - | 183 |