Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039120.2 | NP_001034209.1 | 2 | 327 | ||
GenBank mRNA | BC139131.1 | - | 2 | 327 | 8862153 | |
DQ012037.1 | - | 2 | 283 |