Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039121.1 | NP_001034210.1 | 2 | 109 | ||
GenBank mRNA | BC132224.1 | - | 2 | 93 | 40130501 | |
BC132560.1 | - | 2 | 93 | 40130837 | ||
DQ012045.1 | - | 2 | 109 |