Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039123.1 | NP_001034212.1 | 2 | 161 | ||
GenBank mRNA | DQ012029.1 | - | 2 | 161 |