Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002972.3 | NP_002963.2 | 40 | 5791 | ||
XM_005261935.2 | XP_005261992.1 | 39 | 5661 | |||
XM_011530709.2 | XP_011529011.1 | 40 | 5609 | |||
XM_011530710.2 | XP_011529012.1 | 40 | 5605 | |||
XM_017028905.1 | XP_016884394.1 | 39 | 5530 | |||
GenBank mRNA | BC009268.1 | - | 18 | 2672 | ||
BC087612.1 | - | 40 | 5730 | 6671883 | ||
U93181.1 | - | 33 | 4785 |