Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002972.3 | NP_002963.2 | 7 | 1002 | ||
XM_005261935.2 | XP_005261992.1 | 7 | 950 | |||
XM_011530709.2 | XP_011529011.1 | 7 | 820 | |||
XM_011530710.2 | XP_011529012.1 | 7 | 816 | |||
XM_017028905.1 | XP_016884394.1 | 7 | 819 | |||
GenBank mRNA | AB209682.1 | - | 6 | 690 | ||
AK057985.1 | - | 8 | 827 | |||
BC040031.1 | - | 7 | 953 | |||
BC087612.1 | - | 7 | 941 | 6671883 | ||
CR749672.1 | - | 5 | 618 | |||
U93181.1 | - | 1 | 77 |