Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000332.3 | NP_000323.2 | 9 | 3024 | ||
NM_001128164.1 | NP_001121636.1 | 8 | 2975 | |||
GenBank mRNA | BC113082.1 | - | 5 | 637 | ||
BC117125.1 | - | 2 | 2086 | 40125676 | ||
X79204.1 | - | 9 | 2991 |