Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006919.2 | NP_008850.1 | 8 | 1121 | ||
GenBank mRNA | AB046399.1 | - | 7 | 886 | ||
AF461146.1 | - | 7 | 977 | |||
AJ515706.1 | - | 7 | 1007 | |||
AK222746.1 | - | 8 | 1051 | |||
AK222753.1 | - | 8 | 1051 | |||
AK314805.1 | - | 8 | 1051 | |||
AY190327.1 | - | 5 | 654 | |||
AY245781.1 | - | 7 | 977 | |||
BC005224.1 | - | 8 | 1051 | 4101988 | ||
BC008483.1 | - | 8 | 1051 | |||
BT006748.1 | - | 7 | 977 | |||
EU852041.1 | - | 7 | 977 | |||
S66896.1 | - | 8 | 1038 | |||
U19556.1 | - | 7 | 1006 |