Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008828.3 | NP_032854.2 | 11 | 1545 | ||
GenBank mRNA | AK133877.1 | - | 11 | 1467 | ||
AK145846.1 | - | 11 | 1481 | |||
AK164440.1 | - | 11 | 1467 | |||
AK167441.1 | - | 11 | 1439 | |||
AK167459.1 | - | 11 | 1439 | |||
AK167710.1 | - | 11 | 1438 | |||
AK183989.1 | - | 1 | 72 | |||
AK198977.1 | - | 2 | 208 | |||
AK200341.1 | - | 2 | 207 | |||
AK211448.1 | - | 2 | 289 | |||
AK216492.1 | - | 2 | 288 | |||
AK219796.1 | - | 2 | 207 | |||
BC023819.1 | - | 11 | 1451 | |||
BC032167.1 | - | 11 | 1451 | |||
BC083355.1 | - | 11 | 1437 | 6828087 | ||
BC108372.1 | - | 11 | 1449 | 5060103 |