Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006919.2 | NP_008850.1 | 7 | 865 | ||
GenBank mRNA | AF428135.1 | - | 6 | 721 | ||
AF428136.1 | - | 6 | 721 | |||
AF428137.1 | - | 6 | 721 | |||
AF428138.1 | - | 7 | 721 | |||
AF428139.1 | - | 7 | 721 | |||
AF461146.1 | - | 6 | 721 | |||
AJ515706.1 | - | 6 | 751 | |||
AK222746.1 | - | 7 | 795 | |||
AK222753.1 | - | 7 | 795 | |||
AK314805.1 | - | 7 | 795 | |||
AY190327.1 | - | 4 | 398 | |||
AY245778.1 | - | 6 | 721 | |||
AY245781.1 | - | 6 | 721 | |||
BC005224.1 | - | 7 | 795 | 4101988 | ||
BC008483.1 | - | 7 | 795 | |||
BT006748.1 | - | 6 | 721 | |||
EU852041.1 | - | 6 | 721 | |||
S66896.1 | - | 7 | 782 | |||
U19556.1 | - | 6 | 750 |
Gene Symbol: SERPINB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002974.3 | NP_002965.1 | 7 | 858 | ||
NM_175041.1 | NP_778206.1 | 7 | 795 | |||
XM_011526138.1 | XP_011524440.1 | 7 | 868 | |||
GenBank mRNA | AB046400.1 | - | 6 | 663 | ||
AF461147.1 | - | 6 | 721 | |||
AK292212.1 | - | 7 | 858 | |||
AY245780.1 | - | 6 | 721 | |||
AY245782.1 | - | 6 | 721 | |||
BC017401.1 | - | 7 | 798 | 4047303 | ||
U19557.1 | - | 7 | 771 | |||
X89015.1 | - | 6 | 721 |