Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042388.2 | NP_001035847.1 | 6 | 614 | ||
NM_005134.3 | NP_005125.1 | 6 | 563 | |||
NR_052003.1 | 5 | 456 | ||||
XM_011525775.2 | XP_011524077.1 | 6 | 740 | |||
XM_011525776.1 | XP_011524078.1 | 6 | 604 | |||
XM_011525779.2 | XP_011524081.1 | 6 | 740 | |||
XM_017026105.1 | XP_016881594.1 | 6 | 740 | |||
XM_017026106.1 | XP_016881595.1 | 5 | 418 | |||
XM_017026107.1 | XP_016881596.1 | 5 | 433 | |||
GenBank mRNA | AF111106.1 | - | 6 | 533 | ||
AF200478.1 | - | 6 | 513 | |||
AK027064.1 | - | 6 | 565 | |||
AK292538.1 | - | 6 | 494 | |||
AK293869.1 | - | 5 | 354 | |||
AK315106.1 | - | 6 | 521 | |||
BC060829.1 | - | 6 | 515 | 30342594 | ||
DR006046.1 | - | 6 | 570 |