Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162922.1 | NP_001156394.1 | 4 | 1122 |
Gene Symbol: Gm14326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190302.2 | NP_001177231.1 | 5 | 2028 |