Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110555.1 | NP_001104025.1 | 2 | 1108 | ||
GenBank mRNA | AK036389.1 | - | 2 | 1109 | ||
BC011440.1 | - | 3 | 1183 | 3989862 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097979.2 | NP_001091448.2 | 2 | 1108 |