Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ541076.1 | - | 576 |
Gene Symbol: 1700040F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076905.1 | - | 436 | |||
AK077064.1 | - | 9 | 3098 | |||
FJ541082.1 | - | 513 | ||||
FJ541086.1 | - | 512 | ||||
FJ541088.1 | - | 326 | ||||
FJ541089.1 | - | 378 | ||||
FJ541094.1 | - | 259 | ||||
FJ541095.1 | - | 346 | ||||
FJ541098.1 | - | 310 | ||||
FJ541100.1 | - | 245 | ||||
FJ541102.1 | - | 126 |
Gene Symbol: Pmel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ541090.1 | - | 193 |