Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081172.2 | NP_001074641.1 | 13 | 1288 | ||
GenBank mRNA | AK122409.1 | - | 10 | 914 | ||
BC031840.1 | - | 3 | 148 | |||
BC051097.1 | - | 3 | 148 | |||
BC158060.1 | - | 13 | 1252 | 9007360 |