Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177984.2 | NP_001171455.1 | 15 | 2636 | ||
NM_014191.3 | NP_055006.1 | 15 | 2636 | |||
XM_006719556.3 | XP_006719619.1 | 15 | 3227 | |||
XM_011538650.2 | XP_011536952.1 | 15 | 2655 | |||
XM_011538651.2 | XP_011536953.1 | 15 | 2620 | |||
XM_017019794.1 | XP_016875283.1 | 15 | 2633 | |||
XM_017019795.1 | XP_016875284.1 | 15 | 2652 | |||
XM_017019796.1 | XP_016875285.1 | 15 | 2652 | |||
GenBank mRNA | AB027567.1 | - | 15 | 2636 | ||
AF225988.1 | - | 14 | 2458 | |||
AK309213.1 | - | 15 | 2586 | |||
FJ611941.1 | - | 14 | 2458 |