Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038.5 | NP_001029.1 | 3, 4 | 978 | ||
NM_001159575.1 | NP_001153047.1 | 3, 4 | 849 | |||
NM_001159576.1 | NP_001153048.1 | 2, 3 | 1130 | |||
GenBank mRNA | AK172792.1 | - | 2, 3 | 1133 | ||
AK304379.1 | - | 3, 4 | 851 | |||
BC006526.2 | - | 2, 3 | 816 | |||
BC062613.1 | - | 2, 3 | 745 | 5186683 | ||
DQ402522.1 | - | 2, 3 | 689 | |||
X76180.1 | - | 3, 4 | 788 |