Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110130.1 | NP_001103600.1 | 9 | 553 | ||
GenBank mRNA | AK008394.1 | - | 9 | 531 | ||
BY708305.1 | - | 9 | 530 |