Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099277.1 | NP_001092747.1 | 9, 10 | 3191 | ||
GenBank mRNA | BC157962.1 | - | 7, 8 | 3029 | 9007287 | |
DQ864732.1 | - | 9, 10 | 3191 |