Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011967.3 | NP_036097.1 | 2 | 415 | ||
GenBank mRNA | AF019661.1 | - | 2 | 130 | ||
AK143468.1 | - | 2 | 417 | |||
AK160530.1 | - | 2 | 175 | |||
AK160550.1 | - | 2 | 169 | |||
AK160741.1 | - | 2 | 178 | |||
AK161680.1 | - | 2 | 163 | |||
BC007181.1 | - | 2 | 162 | |||
BC010709.1 | - | 2 | 151 | 2648635 | ||
BC083342.1 | - | 2 | 148 | 5700963 | ||
BC108368.1 | - | 2 | 145 | 3596530 |