Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167151.1 | NP_001160623.1 | 1 | 715 |
Gene Symbol: Gm8453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167150.1 | NP_001160622.1 | 1 | 715 |
Gene Symbol: Gm8693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167154.1 | NP_001160626.1 | 1 | 715 |
Gene Symbol: Vmn1r111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166746.1 | NP_001160218.1 | 1 | 715 |
Gene Symbol: Vmn1r118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166742.1 | NP_001160214.1 | 1 | 715 |
Gene Symbol: Vmn1r131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166839.1 | NP_001160311.1 | 1 | 715 |
Gene Symbol: Vmn1r138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167169.1 | NP_001160641.1 | 1 | 715 |
Gene Symbol: Vmn1r166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167168.1 | NP_001160640.1 | 1 | 715 |