Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL15-CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027921.3 | 3 | 715 | |||
NR_027922.3 | 3 | 715 | ||||
GenBank mRNA | BC140941.1 | - | 3 | 298 | 9056753 |
Gene Symbol: CCL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032965.5 | NP_116741.2 | 3 | 715 | ||
GenBank mRNA | AF031587.1 | - | 3 | 169 | ||
BX497188.1 | - | 3 | 183 | |||
U58914.1 | - | 3 | 229 |