Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204182.1 | NP_001191111.1 | 5 | 860 | ||
NM_001204183.1 | NP_001191112.1 | 5 | 860 | |||
NM_002497.3 | NP_002488.1 | 5 | 860 | |||
XM_005273147.1 | XP_005273204.1 | 5 | 857 | |||
GenBank mRNA | AK223353.1 | - | 5 | 821 | ||
AK225522.1 | - | 5 | 821 | |||
AY045701.1 | - | 5 | 791 | |||
AY863109.1 | - | 1 | 64 | |||
BC043502.1 | - | 5 | 848 | 5541026 | ||
BC052807.1 | - | 5 | 855 | |||
BC065932.1 | - | 5 | 836 | |||
BT019729.1 | - | 5 | 709 | |||
U11050.1 | - | 5 | 843 | |||
Z29066.1 | - | 5 | 791 |