Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igk-V21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S74563.1 | - | 1 | 85 | ||
X67623.1 | - | 1 | 85 | |||
X67625.1 | - | 1 | 85 |