Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042407.1 | NP_001035866.1 | 1, 2 | 145 | ||
GenBank mRNA | AK043682.1 | - | 1, 2 | 119 | ||
BC147304.1 | - | 1, 2 | 130 | 9056372 | ||
BC147305.1 | - | 1, 2 | 130 | 9056374 | ||
CJ064027.1 | - | 1, 2 | 119 | |||
CJ176766.1 | - | 1, 2 | 146 |