Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270537.1 | NP_001257466.1 | 2 | 133 | ||
GenBank mRNA | BY751384.1 | - | 2 | 136 | ||
CA562068.1 | - | 2 | 122 |