Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000035.3 | NP_000026.2 | 7 | 819 | ||
GenBank mRNA | AK026411.1 | - | 7 | 596 | ||
AK290795.1 | - | 7 | 813 | |||
BC029399.1 | - | 7 | 813 | 4593670 | ||
X00270.1 | - | 1 | 26 | |||
X01098.1 | - | 7 | 803 | |||
X02747.1 | - | 7 | 862 |