Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302123.1 | NP_001289052.1 | 3 | 439 | ||
NM_005409.4 | NP_005400.1 | 3 | 439 | |||
GenBank mRNA | AF002985.1 | - | 3 | 316 | ||
AF030514.1 | - | 3 | 298 | |||
AL703031.1 | - | 3 | 279 | |||
BC005292.1 | - | 3 | 289 | 3934139 | ||
BC110986.1 | - | 3 | 298 | 5178174 | ||
BP289021.1 | - | 3 | 341 | |||
BT006787.1 | - | 3 | 229 | |||
U66096.1 | - | 3 | 274 | |||
Y15220.1 | - | 3 | 235 |