Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002994.4 | NP_002985.1 | 2 | 339 | ||
GenBank mRNA | AK312128.1 | - | 2 | 287 | ||
BC008376.1 | - | 2 | 290 | 3826939 | ||
BT006807.1 | - | 2 | 191 | |||
BU169770.1 | - | 2 | 310 | |||
CR457428.1 | - | 2 | 191 | |||
DN992282.1 | - | 2 | 397 | |||
X78686.1 | - | 2 | 297 |