Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177739.3 | NP_808407.2 | 4 | 2398 | ||
GenBank mRNA | AK129283.1 | - | 4 | 2366 | ||
AK162510.1 | - | 2 | 578 | |||
BC050041.1 | - | 4 | 2387 | 30006851 |