Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002998.3 | NP_002989.2 | 5 | 1163 | ||
XM_011517212.2 | XP_011515514.1 | 6 | 695 | |||
GenBank mRNA | AK025488.1 | - | 5 | 1004 | ||
AK074530.1 | - | 6 | 949 | |||
AK097839.1 | - | 6 | 964 | |||
AK130131.1 | - | 5 | 1004 | |||
AK298643.1 | - | 5 | 863 | |||
AK298939.1 | - | 5 | 705 | |||
AK300067.1 | - | 5 | 901 | |||
BC030133.2 | - | 5 | 1149 | 4138627 | ||
BC049836.1 | - | 5 | 881 | 4931549 | ||
J04621.1 | - | 5 | 1133 |