Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002998.3 | NP_002989.2 | 2 | 744 | ||
XM_011517212.2 | XP_011515514.1 | 3 | 276 | |||
GenBank mRNA | AK025488.1 | - | 2 | 585 | ||
AK074530.1 | - | 2 | 530 | |||
AK097839.1 | - | 3 | 545 | |||
AK130131.1 | - | 2 | 585 | |||
AK298643.1 | - | 2 | 444 | |||
BC030133.2 | - | 2 | 730 | 4138627 | ||
BC049836.1 | - | 2 | 462 | 4931549 | ||
J04621.1 | - | 2 | 714 |