Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007067.1 | NP_001007068.1 | 7 | 758 | ||
NM_001007068.1 | NP_001007069.1 | 7 | 743 | |||
NM_001007069.1 | NP_001007070.1 | 7 | 758 | |||
NM_001007070.1 | NP_001007071.1 | 7 | 755 | |||
NM_005625.3 | NP_005616.2 | 7 | 761 | |||
XM_011517571.2 | XP_011515873.2 | 7 | 2130 | |||
XM_017013727.1 | XP_016869216.1 | 7 | 2133 | |||
XM_017013728.1 | XP_016869217.1 | 7 | 2130 | |||
GenBank mRNA | AF000652.1 | - | 7 | 759 | ||
AF006636.1 | - | 7 | 686 | |||
AK128645.1 | - | 8 | 2185 | |||
AK295198.1 | - | 6 | 498 | |||
AK298697.1 | - | 7 | 573 | |||
AK300647.1 | - | 7 | 675 | |||
AK312274.1 | - | 7 | 669 | |||
BC013254.2 | - | 7 | 645 | |||
BC113674.1 | - | 7 | 705 | 8322726 | ||
BC113676.1 | - | 7 | 705 | 8322728 | ||
BC143915.1 | - | 7 | 702 | 9052433 | ||
BC143916.1 | - | 7 | 705 | 9052434 | ||
U83463.1 | - | 6 | 635 |