Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085553.1 | NP_001079022.1 | 1 | 1080 |
Gene Symbol: Gm10922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085554.1 | NP_001079023.1 | 1 | 1080 |
Gene Symbol: Gm14345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085545.1 | NP_001079014.1 | 1 | 1080 |
Gene Symbol: Gm14346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085551.1 | NP_001079020.1 | 1 | 1080 |
Gene Symbol: Gm14347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085543.1 | NP_001079012.1 | 1 | 1080 |
Gene Symbol: Gm14351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085552.2 | NP_001079021.1 | 1 | 1080 |
Gene Symbol: Gm14374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085523.1 | NP_001078992.1 | 1 | 1080 |
Gene Symbol: Gm21950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270667.1 | NP_001257596.1 | 1 | 1035 | ||
GenBank mRNA | AB055855.1 | - | 1 | 1035 |
Gene Symbol: Gm21951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270669.1 | NP_001257598.1 | 1 | 1052 | ||
GenBank mRNA | AB055854.1 | - | 1 | 1052 |
Gene Symbol: Gm2799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168334.1 | NP_001161806.1 | 1 | 1080 |
Gene Symbol: Gm2825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168337.1 | NP_001161809.1 | 1 | 1080 |
Gene Symbol: Gm2863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099333.3 | NP_001092803.3 | 1 | 1080 |
Gene Symbol: Gm2913 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243015.1 | NP_001229944.1 | 1 | 1080 |
Gene Symbol: Gm2927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282034.1 | NP_001268963.1 | 1 | 1080 |
Gene Symbol: Gm2933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145038.1 | NP_001138510.1 | 1 | 1048 | ||
GenBank mRNA | BC115962.1 | - | 1 | 1048 | 40057634 |
Gene Symbol: Gm3701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243011.1 | NP_001229940.1 | 1 | 1080 |
Gene Symbol: Gm3706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243001.1 | NP_001229930.1 | 1 | 1074 |
Gene Symbol: Gm3750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099643.2 | NP_001093113.1 | 1 | 1074 |
Gene Symbol: Gm3763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243024.1 | NP_001229953.1 | 1 | 1080 | ||
GenBank mRNA | BC089532.1 | - | 1 | 1080 | 6704091 |
Gene Symbol: Gmcl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027955.3 | NP_082231.2 | 1 | 1080 | ||
GenBank mRNA | AB055856.1 | - | 1038 | |||
AK006065.1 | - | 1 | 1061 |