Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003000.2 | NP_002991.2 | 4, 5 | 582 | ||
GenBank mRNA | AK312056.1 | - | 4, 5 | 444 | ||
BC007840.2 | - | 4, 5 | 582 | 4124737 | ||
BF796275.1 | - | 4, 5 | 430 | |||
BU554163.1 | - | 2, 3 | 172 | |||
D10245.1 | - | 3, 4 | 377 | |||
DQ403007.1 | - | 4, 5 | 404 | |||
M32246.1 | - | 3, 4 | 377 | |||
U17248.1 | - | 4, 5 | 565 |