Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003000.2 | NP_002991.2 | 3 | 399 | ||
GenBank mRNA | AK312056.1 | - | 3 | 261 | ||
BC007840.2 | - | 3 | 399 | 4124737 | ||
BF796275.1 | - | 3 | 247 | |||
D10245.1 | - | 2 | 194 | |||
DQ403007.1 | - | 3 | 221 | |||
M32246.1 | - | 2 | 194 | |||
U17248.1 | - | 3 | 382 |