Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035511.1 | NP_001030588.1 | 5 | 538 | ||
NM_001035512.1 | NP_001030589.1 | 5 | 600 | |||
NM_001035513.1 | NP_001030590.1 | 4 | 543 | |||
NM_001278172.1 | NP_001265101.1 | 4 | 436 | |||
NM_003001.3 | NP_002992.1 | 6 | 702 | |||
NR_103459.1 | 7 | 729 | ||||
GenBank mRNA | AK131051.1 | - | 6 | 697 | ||
AK293687.1 | - | 5 | 592 | |||
AK294305.1 | - | 5 | 533 | |||
BC020808.1 | - | 6 | 697 | 4291847 | ||
BC033626.1 | - | 6 | 695 | 5416956 | ||
BC066329.1 | - | 6 | 697 | 5301907 | ||
D49737.1 | - | 6 | 685 | |||
U57877.1 | - | 6 | 699 |